Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014831.2 | NP_055646.2 | 8 | 1174 | ||
XM_006713432.3 | XP_006713495.1 | 9 | 1342 | |||
XM_011534289.2 | XP_011532591.1 | 9 | 2103 | |||
XM_017007571.1 | XP_016863060.1 | 9 | 1235 | |||
XM_017007572.1 | XP_016863061.1 | 9 | 1137 | |||
XM_017007573.1 | XP_016863062.1 | 9 | 1145 |