Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252100.1 | NP_001239029.1 | 19, 20 | 3148 | ||
NM_001252102.1 | NP_001239031.1 | 19, 20 | 3148 | |||
NM_001252103.1 | NP_001239032.1 | 19, 20 | 3148 | |||
NM_017596.3 | NP_060066.2 | 19, 20 | 3148 | |||
XM_017000731.1 | XP_016856220.1 | 18, 19 | 2791 | |||
XM_017000732.1 | XP_016856221.1 | 13, 14 | 1932 | |||
GenBank mRNA | AB007918.2 | - | 19, 20 | 3148 | ||
AM177180.1 | - | 19, 20 | 2831 | |||
BC137281.1 | - | 19, 20 | 2856 | 9021278 | ||
BC144557.1 | - | 19, 20 | 2856 | 9053085 |