Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYNRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025081.2 | NP_079357.2 | 4 | 2181 | ||
GenBank mRNA | AB037726.2 | - | 2 | 1626 | ||
BX248773.1 | - | 2 | 1188 |