Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291366.1 | NP_001278295.1 | 3 | 2265 | ||
NM_001291367.1 | NP_001278296.1 | 4 | 1912 | |||
XM_017002583.1 | XP_016858072.1 | 3 | 2367 | |||
XM_017002584.1 | XP_016858073.1 | 3 | 2390 | |||
GenBank mRNA | AK123855.1 | - | 4 | 1888 | ||
AK312221.1 | - | 1 | 21 | |||
BC006300.2 | - | 1 | 62 | |||
KF150175.1 | - | 3 | 2265 |