Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1495 | |||
NR_136193.1 | 4 | 2192 | ||||
NR_136194.1 | 4 | 1662 | ||||
NR_136197.1 | 4 | 1566 | ||||
NR_136198.1 | 3 | 1383 | ||||
NR_136199.1 | 5 | 1705 | ||||
NR_136200.1 | 4 | 2195 | ||||
NR_136201.1 | 4 | 2362 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 2103 | ||
AK021601.1 | - | 1 | 1842 | |||
DQ831955.1 | - | 1 | 255 | |||
DQ831956.1 | - | 1 | 255 | |||
EF529495.1 | - | 4 | 495 | |||
EF529498.1 | - | 4 | 630 | |||
EF529506.1 | - | 4 | 495 | |||
EF529508.1 | - | 4 | 495 | |||
EU000853.1 | - | 4 | 1408 | |||
EU000854.1 | - | 4 | 1408 | |||
EU000855.1 | - | 3 | 2500 | |||
EU000856.1 | - | 3 | 2500 |