Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243342.1 | NP_001230271.1 | 11 | 1668 | ||
NM_017950.3 | NP_060420.2 | 11 | 1668 | |||
XM_011524963.2 | XP_011523265.1 | 11 | 1705 | |||
XM_011524964.2 | XP_011523266.1 | 5 | 637 | |||
XM_011524965.2 | XP_011523267.1 | 11 | 1668 | |||
XM_017024807.1 | XP_016880296.1 | 11 | 1668 | |||
GenBank mRNA | AB046860.1 | - | 7 | 1174 | ||
AL713758.1 | - | 1 | 178 | |||
BC035251.1 | - | 11 | 1661 | 4838111 |