Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 23 | 4064 | ||
XM_005261909.1 | XP_005261966.1 | 24 | 4112 | |||
XM_005261910.2 | XP_005261967.1 | 24 | 4123 | |||
XM_005261911.4 | XP_005261968.1 | 23 | 4062 | |||
XM_006724413.2 | XP_006724476.1 | 25 | 4182 | |||
XM_011530682.2 | XP_011528984.1 | 24 | 4263 | |||
XM_017028703.1 | XP_016884192.1 | 25 | 4163 | |||
XM_017028704.1 | XP_016884193.1 | 24 | 4116 | |||
GenBank mRNA | AB002313.1 | - | 22 | 3981 | ||
AB075511.1 | - | 8 | 1174 | |||
AK025701.1 | - | 10 | 1432 | |||
AK056543.1 | - | 6 | 1233 | |||
AK074932.1 | - | 5 | 700 | |||
AK126394.1 | - | 4 | 773 | |||
BC148225.1 | - | 1 | 243 | |||
BC148280.1 | - | 1 | 243 | |||
S76730.1 | - | 1 | 216 |