Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 31 | 5114 | ||
XM_005261909.1 | XP_005261966.1 | 32 | 5162 | |||
XM_005261910.2 | XP_005261967.1 | 32 | 5173 | |||
XM_005261911.4 | XP_005261968.1 | 31 | 5112 | |||
XM_006724413.2 | XP_006724476.1 | 33 | 5232 | |||
XM_011530682.2 | XP_011528984.1 | 32 | 5313 | |||
XM_017028703.1 | XP_016884192.1 | 33 | 5213 | |||
XM_017028704.1 | XP_016884193.1 | 32 | 5166 | |||
GenBank mRNA | AB002313.1 | - | 30 | 5031 | ||
AB075511.1 | - | 16 | 2224 | |||
AK025415.1 | - | 6 | 701 | |||
AK025701.1 | - | 18 | 2482 | |||
AK056543.1 | - | 14 | 2283 | |||
AK074932.1 | - | 13 | 1750 | |||
AK123131.1 | - | 3 | 695 | |||
AK126394.1 | - | 12 | 1823 | |||
BC004542.2 | - | 8 | 933 | |||
BC110366.1 | - | 3 | 351 | |||
BT006887.1 | - | 8 | 917 | |||
S76730.1 | - | 9 | 1263 |