Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 11 | 1145 | ||
XM_005273409.1 | XP_005273466.1 | 11 | 1145 | |||
XM_005273410.1 | XP_005273467.1 | 11 | 1145 | |||
XM_005273411.1 | XP_005273468.1 | 9 | 953 | |||
XM_005273412.3 | XP_005273469.1 | 11 | 1145 | |||
XM_017013088.1 | XP_016868577.1 | 9 | 953 | |||
XM_017013089.1 | XP_016868578.1 | 10 | 1018 | |||
GenBank mRNA | AK057293.1 | - | 2 | 535 | ||
AK304441.1 | - | 11 | 1097 | |||
CR749838.1 | - | 7 | 569 |