Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 16 | 1621 | ||
XM_005273409.1 | XP_005273466.1 | 17 | 1732 | |||
XM_005273410.1 | XP_005273467.1 | 17 | 1708 | |||
XM_005273411.1 | XP_005273468.1 | 15 | 1540 | |||
XM_017013088.1 | XP_016868577.1 | 14 | 1429 | |||
XM_017013089.1 | XP_016868578.1 | 15 | 1494 | |||
GenBank mRNA | AK057293.1 | - | 7 | 1011 | ||
AK074839.1 | - | 3 | 191 | |||
AK304441.1 | - | 16 | 1573 | |||
AK308922.1 | - | 2 | 889 | |||
CR749838.1 | - | 12 | 1045 |