Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144708.3 | NP_653309.3 | 2 | 393 | ||
XM_011510673.2 | XP_011508975.1 | 2 | 879 | |||
XM_011510674.1 | XP_011508976.1 | 2 | 879 | |||
XM_011510675.2 | XP_011508977.1 | 2 | 879 | |||
XM_011510676.2 | XP_011508978.1 | 2 | 879 | |||
XM_011510677.2 | XP_011508979.1 | 2 | 879 | |||
XM_011510679.2 | XP_011508981.1 | 2 | 879 | |||
XM_011510680.1 | XP_011508982.1 | 2 | 879 | |||
XM_011510681.1 | XP_011508983.1 | 2 | 879 | |||
XM_011510682.2 | XP_011508984.1 | 2 | 879 | |||
XM_011510686.2 | XP_011508988.1 | 2 | 879 | |||
XM_011510688.1 | XP_011508990.1 | 2 | 879 | |||
XM_011510689.2 | XP_011508991.1 | 2 | 879 | |||
XM_017003413.1 | XP_016858902.1 | 2 | 879 | |||
XM_017003414.1 | XP_016858903.1 | 2 | 879 | |||
XM_017003417.1 | XP_016858906.1 | 2 | 879 | |||
GenBank mRNA | AJ549812.1 | - | 1 | 92 | ||
AK126073.1 | - | 2 | 393 |