Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015312.3 | NP_056127.2 | 5 | 492 | ||
XM_005263282.1 | XP_005263339.1 | 7 | 820 | |||
XM_005263287.1 | XP_005263344.1 | 7 | 820 | |||
XM_006714344.1 | XP_006714407.1 | 7 | 820 | |||
XM_011532319.1 | XP_011530621.1 | 7 | 820 | |||
XM_011532320.2 | XP_011530622.1 | 8 | 915 | |||
XM_011532321.2 | XP_011530623.1 | 6 | 622 | |||
XM_011532322.1 | XP_011530624.1 | 7 | 820 | |||
XM_011532323.1 | XP_011530625.1 | 7 | 820 | |||
XM_011532324.1 | XP_011530626.1 | 7 | 820 | |||
XM_011532325.1 | XP_011530627.1 | 7 | 820 | |||
XM_011532326.1 | XP_011530628.1 | 7 | 820 | |||
XM_011532327.1 | XP_011530629.1 | 8 | 866 | |||
XM_017008695.1 | XP_016864184.1 | 7 | 820 | |||
XM_017008696.1 | XP_016864185.1 | 7 | 820 | |||
XM_017008697.1 | XP_016864186.1 | 7 | 820 | |||
XM_017008699.1 | XP_016864188.1 | 7 | 820 | |||
GenBank mRNA | DQ335469.1 | - | 5 | 492 |