Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291088.1 | NP_001278017.1 | 4 | 3291 | ||
NM_031951.4 | NP_114157.4 | 4 | 3174 | |||
XM_011527360.2 | XP_011525662.1 | 4 | 3118 | |||
GenBank mRNA | AK128826.1 | - | 4 | 3145 | ||
AK302913.1 | - | 4 | 3183 | |||
AY026504.2 | - | 4 | 3202 |