Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 11 | 1903 | ||
NM_001136030.2 | NP_001129502.1 | 11 | 2000 | |||
NM_001261844.1 | NP_001248773.1 | 10 | 1752 | |||
NM_014796.2 | NP_055611.1 | 9 | 1843 | |||
XM_005269247.2 | XP_005269304.1 | 10 | 1826 | |||
XM_006719715.3 | XP_006719778.1 | 13 | 2334 | |||
XM_011539035.2 | XP_011537337.1 | 13 | 2347 | |||
XM_011539036.1 | XP_011537338.1 | 12 | 2168 | |||
XM_011539037.2 | XP_011537339.1 | 11 | 6388 | |||
XM_017020262.1 | XP_016875751.1 | 13 | 3397 | |||
XM_017020263.1 | XP_016875752.1 | 13 | 3303 | |||
GenBank mRNA | AB018291.1 | - | 9 | 2041 | ||
AF161375.1 | - | 3 | 748 | |||
AK295026.1 | - | 9 | 1647 | |||
AK298405.1 | - | 9 | 1669 | |||
AK303223.1 | - | 11 | 1926 | |||
BC044578.1 | - | 9 | 1671 | 5275264 |