Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 11 | 2265 | ||
NM_001136030.2 | NP_001129502.1 | 11 | 2362 | |||
NM_001261844.1 | NP_001248773.1 | 10 | 2114 | |||
NM_014796.2 | NP_055611.1 | 9 | 2205 | |||
XM_005269247.2 | XP_005269304.1 | 10 | 2188 | |||
XM_006719715.3 | XP_006719778.1 | 13 | 2696 | |||
XM_011539035.2 | XP_011537337.1 | 13 | 2709 | |||
XM_011539036.1 | XP_011537338.1 | 12 | 2530 | |||
XM_011539037.2 | XP_011537339.1 | 11 | 6750 | |||
XM_017020262.1 | XP_016875751.1 | 13 | 3759 | |||
XM_017020263.1 | XP_016875752.1 | 13 | 3665 | |||
GenBank mRNA | AB018291.1 | - | 9 | 2403 |