Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 19 | 1914 | ||
XM_011545742.2 | XP_011544044.1 | 22 | 2124 | |||
XM_011545744.2 | XP_011544046.1 | 7 | 724 | |||
XM_011545745.1 | XP_011544047.1 | 5 | 492 | |||
XM_011545746.2 | XP_011544048.1 | 5 | 595 | |||
XM_017022947.1 | XP_016878436.1 | 23 | 2197 | |||
XM_017022948.1 | XP_016878437.1 | 6 | 1296 | |||
XM_017022949.1 | XP_016878438.1 | 4 | 1567 | |||
GenBank mRNA | AK098260.1 | - | 17 | 1871 | ||
AK128606.1 | - | 8 | 782 | |||
AK307544.1 | - | 20 | 1973 | |||
BC109294.1 | - | 7 | 642 | |||
BC109295.1 | - | 7 | 642 |