Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 34 | 4023 | ||
XM_011545742.2 | XP_011544044.1 | 38 | 4305 | |||
XM_011545744.2 | XP_011544046.1 | 23 | 2905 | |||
XM_011545745.1 | XP_011544047.1 | 21 | 2673 | |||
XM_011545746.2 | XP_011544048.1 | 21 | 2776 | |||
XM_017022947.1 | XP_016878436.1 | 39 | 4378 | |||
XM_017022948.1 | XP_016878437.1 | 22 | 3477 | |||
XM_017022949.1 | XP_016878438.1 | 20 | 3748 | |||
GenBank mRNA | AK093467.1 | - | 10 | 1586 | ||
AK128606.1 | - | 23 | 2891 | |||
BC038400.1 | - | 11 | 1468 |