Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015016.1 | NP_055831.1 | 27 | 4881 | ||
XM_005259823.3 | XP_005259880.1 | 27 | 5272 | |||
XM_005259824.2 | XP_005259881.1 | 28 | 4958 | |||
XM_005259825.2 | XP_005259882.1 | 28 | 4955 | |||
XM_005259826.3 | XP_005259883.1 | 26 | 5068 | |||
XM_006722698.2 | XP_006722761.1 | 27 | 4950 | |||
XM_006722699.2 | XP_006722762.2 | 27 | 5259 | |||
XM_006722700.3 | XP_006722763.1 | 26 | 5041 | |||
XM_011527823.1 | XP_011526125.1 | 29 | 5048 | |||
XM_017026506.1 | XP_016881995.1 | 27 | 5256 | |||
XM_017026507.1 | XP_016881996.1 | 26 | 5238 | |||
XM_017026508.1 | XP_016881997.1 | 26 | 5236 | |||
XM_017026509.1 | XP_016881998.1 | 27 | 5232 | |||
XM_017026511.1 | XP_016882000.1 | 28 | 5346 | |||
XM_017026512.1 | XP_016882001.1 | 27 | 4937 | |||
XM_017026513.1 | XP_016882002.1 | 27 | 4934 | |||
XM_017026514.1 | XP_016882003.1 | 28 | 4941 | |||
XM_017026515.1 | XP_016882004.1 | 27 | 4947 | |||
XM_017026516.1 | XP_016882005.1 | 27 | 5229 | |||
GenBank mRNA | AB011133.1 | - | 27 | 4880 |