Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080400.1 | NP_001073869.1 | 3 | 1336 | ||
XM_006722866.2 | XP_006722929.1 | 5 | 3297 | |||
XM_006722868.3 | XP_006722931.1 | 5 | 3293 | |||
XM_011528233.2 | XP_011526535.1 | 6 | 3479 | |||
XM_017027192.1 | XP_016882681.1 | 6 | 3482 | |||
XM_017027193.1 | XP_016882682.1 | 6 | 3429 | |||
XM_017027194.1 | XP_016882683.1 | 6 | 3464 | |||
GenBank mRNA | AB067468.1 | - | 1112 |