Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085487.2 | NP_001078956.1 | 8 | 936 | ||
XM_006710314.3 | XP_006710377.1 | 8 | 952 | |||
XM_011540573.2 | XP_011538875.1 | 9 | 916 | |||
XM_011540574.2 | XP_011538876.1 | 8 | 1184 | |||
XM_017000195.1 | XP_016855684.1 | 9 | 988 | |||
XM_017000196.1 | XP_016855685.1 | 6 | 838 | |||
XM_017000197.1 | XP_016855686.1 | 4 | 616 | |||
GenBank mRNA | AB067502.1 | - | 4 | 962 | ||
AK126835.1 | - | 2 | 485 | |||
BX648168.1 | - | 5 | 672 |