Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM69C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044369.2 | NP_001037834.2 | 4 | 1598 | ||
XM_011525816.2 | XP_011524118.1 | 4 | 1644 | |||
XM_017025551.1 | XP_016881040.1 | 4 | 1524 | |||
GenBank mRNA | BC019628.1 | - | 3 | 885 |