Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136263.1 | NP_001129735.1 | 2 | 2550 | ||
XM_011527694.1 | XP_011525996.1 | 3 | 1961 | |||
GenBank mRNA | AB075837.1 | - | 2 | 2550 | ||
AL834341.1 | - | 1 | 2298 |