Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC4H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178032.2 | NP_001171503.1 | 2 | 434 | ||
NM_001178033.2 | NP_001171504.1 | 2 | 272 | |||
NM_001243804.1 | NP_001230733.1 | 2 | 179 | |||
NM_018684.3 | NP_061154.1 | 2 | 272 | |||
NR_045044.1 | 2 | 527 | ||||
GenBank mRNA | AB032992.1 | - | 2 | 272 | ||
AF270491.1 | - | 2 | 204 | |||
AK022807.1 | - | 2 | 403 | |||
AK022918.1 | - | 2 | 166 | |||
AK123763.1 | - | 3 | 665 | |||
AK222708.1 | - | 2 | 189 | |||
AK293570.1 | - | 2 | 189 | |||
BC004411.1 | - | 2 | 223 | 3639978 | ||
BI770655.1 | - | 2 | 356 | |||
BP376634.1 | - | 2 | 179 | |||
CX782941.1 | - | 2 | 144 | |||
DA037517.1 | - | 2 | 473 |