Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284301.2 | NP_001271230.1 | 3 | 617 | ||
NM_001284302.2 | NP_001271231.1 | 3 | 617 | |||
NM_018685.4 | NP_061155.2 | 3 | 617 | |||
XM_006715746.1 | XP_006715809.1 | 3 | 614 | |||
XM_006715747.3 | XP_006715810.1 | 3 | 614 | |||
XM_017012354.1 | XP_016867843.1 | 3 | 614 | |||
XM_017012355.1 | XP_016867844.1 | 3 | 614 | |||
XM_017012356.1 | XP_016867845.1 | 3 | 614 | |||
GenBank mRNA | AF273437.1 | - | 3 | 549 | ||
AK291254.1 | - | 3 | 549 | |||
BC070066.1 | - | 3 | 599 | 4825802 | ||
BQ434804.1 | - | 3 | 597 |