Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018686.5 | NP_061156.1 | 3, 4 | 694 | ||
NR_135117.1 | 3, 4 | 694 | ||||
GenBank mRNA | AF271388.1 | - | 3, 4 | 582 | ||
AF397212.1 | - | 3, 4 | 564 | |||
AK022927.1 | - | 3, 4 | 604 | |||
AK222981.1 | - | 3, 4 | 593 | |||
AL832975.1 | - | 3, 4 | 642 | |||
BC016609.1 | - | 3, 4 | 643 | 3448277 |