Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018688.4 | NP_061158.1 | 7 | 522 | ||
XM_005273573.3 | XP_005273630.1 | 6 | 428 | |||
XM_011544586.2 | XP_011542888.1 | 8 | 607 | |||
GenBank mRNA | AF271732.1 | - | 7 | 485 | ||
AK001289.1 | - | 5 | 384 | |||
AK023980.1 | - | 7 | 469 | |||
AK124486.1 | - | 1 | 807 | |||
AK222470.1 | - | 7 | 494 | |||
BC001223.1 | - | 6 | 406 | 3534031 | ||
BC009824.2 | - | 7 | 456 | 3953604 |