Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205280.1 | NP_001192209.1 | 5 | 661 | ||
XM_017026319.1 | XP_016881808.1 | 4 | 770 | |||
XM_017026323.1 | XP_016881812.1 | 5 | 835 | |||
XM_017026324.1 | XP_016881813.1 | 3 | 616 | |||
XM_017026325.1 | XP_016881814.1 | 4 | 681 | |||
GenBank mRNA | BG197674.1 | - | 2 | 105 | ||
BX112065.1 | - | 5 | 661 |