Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168241.1 | NP_001161713.1 | 6 | 2613 | ||
NM_001191033.1 | NP_001177962.1 | 6 | 3609 | |||
XM_006711951.3 | XP_006712014.1 | 7 | 2170 | |||
XM_011532564.2 | XP_011530866.1 | 6 | 3050 | |||
XM_011532565.2 | XP_011530867.1 | 7 | 2623 | |||
XM_011532566.2 | XP_011530868.1 | 6 | 2381 | |||
GenBank mRNA | AB015349.1 | - | 3 | 668 | ||
AB124552.1 | - | 6 | 2613 | |||
AK090454.1 | - | 6 | 3611 |