Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099777.3 | NP_001093247.1 | 4 | 480 | ||
NM_001130002.2 | NP_001123474.1 | 4 | 480 | |||
XM_017005754.1 | XP_016861243.1 | 4 | 434 | |||
GenBank mRNA | BC029129.1 | - | 4 | 427 | ||
BC126187.1 | - | 4 | 447 | 8991903 | ||
BC126189.1 | - | 4 | 447 | 8991905 |