Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099777.3 | NP_001093247.1 | 5 | 901 | ||
NM_001243729.1 | NP_001230658.1 | 4 | 812 | |||
XM_017005754.1 | XP_016861243.1 | 6 | 951 | |||
GenBank mRNA | BC126187.1 | - | 5 | 866 | 8991903 | |
BC126189.1 | - | 5 | 866 | 8991905 | ||
BC144066.1 | - | 4 | 777 | |||
BG209594.1 | - | 1 | 403 | |||
BG217991.1 | - | 3 | 646 |