Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017922.1 | NP_001017922.1 | 12 | 2236 | ||
NM_018538.3 | NP_061008.2 | 11 | 2132 | |||
XM_006710313.3 | XP_006710376.1 | 13 | 2501 | |||
XM_011540570.2 | XP_011538872.1 | 12 | 2192 | |||
XM_011540571.2 | XP_011538873.1 | 11 | 2199 | |||
GenBank mRNA | AF119899.1 | - | 1 | 98 | ||
AF311285.1 | - | 11 | 2132 | |||
AK056138.1 | - | 12 | 2211 | |||
AK125447.1 | - | 11 | 1980 | |||
BX537371.1 | - | 9 | 2735 |