Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130699.1 | 2 | 252 | |||
NR_130700.1 | 2 | 252 | ||||
NR_130701.1 | 2 | 252 | ||||
NR_130702.1 | 2 | 252 | ||||
GenBank mRNA | AK125248.1 | - | 2 | 189 | ||
AK298283.1 | - | 2 | 192 | |||
AK307199.1 | - | 2 | 192 | |||
AK309231.1 | - | 2 | 201 | |||
BC090920.1 | - | 1 | 83 | |||
DA474648.1 | - | 2 | 252 |