Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277372.2 | NP_001264301.2 | 7, 8 | 1538 | ||
XM_006712172.2 | XP_006712235.1 | 7, 8 | 1546 | |||
XM_011510467.2 | XP_011508769.1 | 7, 8 | 1546 | |||
XM_011510468.2 | XP_011508770.1 | 7, 8 | 1546 | |||
XM_017003111.1 | XP_016858600.1 | 7, 8 | 1546 | |||
XM_017003112.1 | XP_016858601.1 | 7, 8 | 1546 | |||
GenBank mRNA | AK096380.1 | - | 8, 9 | 1645 | ||
AK295603.1 | - | 7, 8 | 1502 | |||
AK297413.1 | - | 7, 8 | 1536 |