Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288742.1 | NP_001275671.1 | 5 | 470 | ||
NM_001288743.1 | NP_001275672.1 | 7 | 658 | |||
XM_011539383.2 | XP_011537685.1 | 6 | 681 | |||
GenBank mRNA | AL832399.1 | - | 1 | 533 | ||
BU145920.1 | - | 5 | 382 |