Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018557.2 | NP_061027.2 | 24 | 4795 | ||
XM_017004341.1 | XP_016859830.1 | 24 | 9371 | |||
GenBank mRNA | AF176832.1 | - | 24 | 4795 | ||
AK054663.1 | - | 9 | 1537 |