Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290268.1 | NP_001277197.1 | 22 | 3926 | ||
NM_080829.3 | NP_543019.2 | 22 | 3859 | |||
NR_110890.1 | 22 | 4085 | ||||
XM_005260294.3 | XP_005260351.1 | 23 | 3923 | |||
XM_006723713.3 | XP_006723776.1 | 22 | 3971 | |||
XM_011528578.2 | XP_011526880.1 | 23 | 3951 | |||
XM_011528579.2 | XP_011526881.1 | 23 | 4089 | |||
XM_011528580.2 | XP_011526882.1 | 23 | 4143 | |||
XM_011528581.2 | XP_011526883.1 | 23 | 4127 | |||
XM_011528585.2 | XP_011526887.1 | 12 | 3274 | |||
XM_017027681.1 | XP_016883170.1 | 14 | 2919 | |||
GenBank mRNA | AV703941.1 | - | 1 | 228 | ||
BX111440.1 | - | 1 | 544 |