Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136108.2 | NP_001129580.2 | 3 | 258 | ||
NM_001301650.1 | NP_001288579.1 | 3 | 258 | |||
NR_125897.1 | 3 | 258 | ||||
GenBank mRNA | AK293465.1 | - | 3 | 242 | ||
AL050297.1 | - | 3 | 236 | |||
BC032421.1 | - | 3 | 225 | |||
BC050572.1 | - | 2 | 197 | |||
BC143822.1 | - | 3 | 442 | 9052337 | ||
BC143827.1 | - | 3 | 442 | |||
BC143829.1 | - | 3 | 442 | 9052344 | ||
BX426634.2 | - | 3 | 271 |