Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002936.2 | 1 | 652 | |||
GenBank mRNA | AK292767.1 | - | 1 | 609 | ||
BC015543.1 | - | 1 | 672 | |||
BC039027.1 | - | 1 | 690 | |||
BC050562.1 | - | 1 | 652 | |||
BC126996.1 | - | 1 | 214 | |||
BC126997.1 | - | 1 | 214 | |||
HQ268503.1 | - | 1 | 214 |