Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017395.1 | XP_016872884.1 | 20 | 4707 | ||
GenBank mRNA | AB028953.1 | - | 8 | 2342 |