Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162499.1 | NP_001155971.1 | 10 | 2943 | ||
NM_012298.2 | NP_036430.1 | 8 | 2664 | |||
XM_011533503.2 | XP_011531805.1 | 10 | 2961 | |||
XM_011533504.2 | XP_011531806.1 | 10 | 3057 | |||
GenBank mRNA | AB014567.1 | - | 8 | 2592 | ||
BC136592.1 | - | 10 | 2882 | 9020582 | ||
BX502783.1 | - | 1 | 578 | |||
GU727621.1 | - | 10 | 2943 |