Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080424.1 | NP_001073893.1 | 11 | 3120 | ||
XM_005256549.3 | XP_005256606.1 | 12 | 3272 | |||
XM_005256550.4 | XP_005256607.1 | 11 | 3114 | |||
XM_005256551.3 | XP_005256608.1 | 11 | 3153 | |||
XM_005256552.4 | XP_005256609.1 | 11 | 4126 | |||
XM_006721483.3 | XP_006721546.1 | 12 | 3217 | |||
XM_011523750.2 | XP_011522052.1 | 12 | 4477 | |||
XM_011523752.2 | XP_011522054.1 | 11 | 3117 | |||
XM_017024381.1 | XP_016879870.1 | 11 | 4563 | |||
GenBank mRNA | AB002344.2 | - | 11 | 3125 |