Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015131.2 | NP_055946.1 | 2 | 368 | ||
GenBank mRNA | AK000836.1 | - | 1 | 64 | ||
BC033206.1 | - | 1 | 73 | |||
D26488.1 | - | 2 | 341 | |||
DA654485.1 | - | 2 | 368 | |||
DA943836.1 | - | 2 | 344 |