Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278557.1 | NP_001265486.1 | 9 | 2088 | ||
NM_001278558.1 | NP_001265487.1 | 7 | 1811 | |||
NM_018639.4 | NP_061109.1 | 9 | 2188 | |||
XM_017019642.1 | XP_016875131.1 | 9 | 2090 | |||
GenBank mRNA | AF038187.1 | - | 4 | 1317 | ||
AF163324.1 | - | 8 | 2026 | |||
AF229181.1 | - | 9 | 2127 | |||
BC015887.2 | - | 9 | 2075 | 3910968 |