Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242590.2 | NP_001229519.1 | 3 | 698 | ||
NM_018643.4 | NP_061113.1 | 4 | 891 | |||
NR_136332.1 | 4 | 891 | ||||
XM_017010956.1 | XP_016866445.1 | 4 | 831 | |||
XM_017010957.1 | XP_016866446.1 | 3 | 635 | |||
GenBank mRNA | AF196329.1 | - | 4 | 848 | ||
AF287008.1 | - | 4 | 865 | |||
AK292162.1 | - | 4 | 828 | |||
AK301519.1 | - | 3 | 635 | |||
BC017773.1 | - | 4 | 846 | 4692680 | ||
DA362458.1 | - | 3 | 338 |