Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 15 | 2548 | ||
GenBank mRNA | AF365927.1 | - | 15 | 2535 | ||
AF365928.1 | - | 11 | 1962 | |||
AJ243500.2 | - | 15 | 2518 | |||
AJ243501.2 | - | 15 | 2518 | |||
AJ487964.2 | - | 16 | 2488 | |||
AK075103.1 | - | 9 | 2049 | |||
AK075206.1 | - | 5 | 918 | |||
AY203947.1 | - | 12 | 2730 | |||
BC034814.1 | - | 1 | 190 | |||
KF534785.1 | - | 15 | 2547 | |||
U82613.1 | - | 1 | 370 | |||
X83877.1 | - | 1 | 370 |