Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046159.2 | 6 | 724 | |||
NR_046160.2 | 6 | 886 | ||||
NR_132339.1 | 5 | 962 | ||||
NR_132340.1 | 5 | 811 | ||||
NR_132341.1 | 5 | 834 | ||||
GenBank mRNA | AK055439.1 | - | 5 | 802 | ||
BC029844.1 | - | 6 | 724 | |||
BC107595.1 | - | 5 | 648 | |||
BC118985.1 | - | 6 | 723 | |||
BC122534.1 | - | 5 | 671 |