Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046159.2 | 6 | 938 | |||
NR_046160.2 | 6 | 1100 | ||||
NR_132339.1 | 5 | 1176 | ||||
NR_132340.1 | 5 | 1025 | ||||
NR_132341.1 | 5 | 1048 | ||||
GenBank mRNA | AK055439.1 | - | 5 | 1016 | ||
BC029844.1 | - | 6 | 938 | |||
BC107595.1 | - | 5 | 862 | |||
BC118985.1 | - | 6 | 937 | |||
BC122534.1 | - | 5 | 885 |