Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527896.2 | XP_011526198.1 | 3 | 323 | ||
XM_017026567.1 | XP_016882056.1 | 2 | 411 |