Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 5 | 480 | ||
XM_005248480.2 | XP_005248537.1 | 5 | 615 | |||
XM_011543306.2 | XP_011541608.1 | 5 | 702 | |||
XM_011543307.2 | XP_011541609.1 | 5 | 1131 | |||
GenBank mRNA | AB020695.1 | - | 5 | 432 | ||
AK290004.1 | - | 5 | 480 | |||
CR936791.1 | - | 6 | 662 | |||
DA326879.1 | - | 5 | 480 |