Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 6 | 699 | ||
NR_046462.1 | 5 | 662 | ||||
XM_005248480.2 | XP_005248537.1 | 6 | 834 | |||
XM_011543306.2 | XP_011541608.1 | 6 | 921 | |||
XM_011543307.2 | XP_011541609.1 | 6 | 1350 | |||
XM_017009332.1 | XP_016864821.1 | 4 | 1597 | |||
XM_017009333.1 | XP_016864822.1 | 4 | 571 | |||
GenBank mRNA | AB020695.1 | - | 6 | 651 | ||
AK290004.1 | - | 6 | 699 | |||
BX640902.1 | - | 5 | 480 | |||
BX647536.1 | - | 5 | 664 |